Assay Details
| Gene Symbol: COX14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257134.1 | NP_001244063.1 | 1 - 2 | 519 | 61 | |
| GenBank mRNA | BG829783.1 | - | 1 - 2 | 253 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257134.1 transcripts.
Set Membership: |
|

COX14
Genomic Map