Assay Details
| Gene Symbol: GZMM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258351.1 | NP_001245280.1 | 2 - 3 | 245 | 79 | |
| NM_005317.3 | NP_005308.1 | 2 - 3 | 249 | 79 | ||
| GenBank mRNA | BC025701.1 | - | 2 - 3 | 221 | 5222281 | 79 |
| CR991101.1 | - | 2 - 3 | 245 | 79 | ||
| L23134.1 | - | 2 - 3 | 211 | 79 | ||
More Information
Set Membership: |
|

GZMM
Genomic Map