Assay Details
| Gene Symbol: G6PC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000151.3 | NP_000142.2 | 1 - 2 | 305 | 80 | |
| NM_001270397.1 | NP_001257326.1 | 1 - 2 | 305 | 80 | ||
| GenBank mRNA | AK303771.1 | - | 1 - 2 | 305 | 80 | |
| AK313982.1 | - | 1 - 2 | 300 | 80 | ||
| BC130478.1 | - | 1 - 2 | 271 | 40146509 | 80 | |
| BC136369.1 | - | 1 - 2 | 271 | 9020356 | 80 | |
| U01120.1 | - | 1 - 2 | 304 | 80 | ||
More Information
Set Membership: |
|

G6PC
Genomic Map