Assay Details
| Gene Symbol: G6PC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270397.1 | NP_001257326.1 | 3 - 4 | 450 | 72 | |
| GenBank mRNA | AK303771.1 | - | 3 - 4 | 450 | 72 | |
More Information
Set Membership: |
|

G6PC
Genomic Map