Assay Details
| Gene Symbol: LIN9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270409.1 | NP_001257338.1 | 11 - 12 | 1504 | 110 | |
| NM_001270410.1 | NP_001257339.1 | 12 - 13 | 1337 | 110 | ||
| NM_173083.3 | NP_775106.2 | 12 - 13 | 1609 | 110 | ||
| XM_005273102.3 | XP_005273159.1 | 12 - 13 | 1668 | 110 | ||
| XM_006711766.2 | XP_006711829.2 | 11 - 12 | 1426 | 110 | ||
| XM_006711767.3 | XP_006711830.1 | 12 - 13 | 1264 | 110 | ||
| XM_017001084.1 | XP_016856573.1 | 10 - 11 | 1464 | 110 | ||
| GenBank mRNA | AF190323.1 | - | 11 - 12 | 1173 | 110 | |
| AF190324.1 | - | 12 - 13 | 1278 | 110 | ||
| AK125690.1 | - | 6 - 7 | 614 | 110 | ||
| AK126177.1 | - | 11 - 12 | 1165 | 110 | ||
| AK308823.1 | - | 11 - 12 | 1255 | 110 | ||
| AY786184.1 | - | 12 - 13 | 1245 | 110 | ||
| BC043444.2 | - | 12 - 13 | 1248 | 110 | ||
| BC045625.1 | - | 12 - 13 | 1603 | 4837217 | 110 | |
| BC065302.1 | - | 12 - 13 | 1323 | 110 | ||
| BX537869.1 | - | 12 - 13 | 1269 | 110 | ||
More Information
Set Membership: |
|

LIN9
Genomic Map