Assay Details
| Gene Symbol: SPATA33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271907.1 | NP_001258836.1 | 1 - 2 | 138 | 77 | |
| NM_001271910.1 | NP_001258839.1 | 1 - 2 | 138 | 77 | ||
| XM_006721133.2 | XP_006721196.1 | 1 - 2 | 138 | 77 | ||
| GenBank mRNA | AK303024.1 | - | 1 - 2 | 104 | 77 | |
| DB452323.1 | - | 1 - 2 | 138 | 77 | ||
| DC357681.1 | - | 1 - 2 | 96 | 77 | ||
| DC374690.1 | - | 1 - 2 | 106 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK303024.1, NM_001271907.1, NM_001271910.1, XM_006721133.2 transcripts.
Set Membership: |
|

SPATA33
Genomic Map