Assay Details
| Gene Symbol: SLC39A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271957.1 | NP_001258886.1 | 2 - 3 | 532 | 72 | |
| NM_001271958.1 | NP_001258887.1 | 2 - 3 | 369 | 72 | ||
| NM_001271959.1 | NP_001258888.1 | 2 - 3 | 538 | 72 | ||
| NM_001271960.1 | NP_001258889.1 | 3 - 4 | 430 | 72 | ||
| NM_014437.4 | NP_055252.2 | 3 - 4 | 676 | 72 | ||
| GenBank mRNA | AF132942.1 | - | 1 - 2 | 226 | 72 | |
| AF151829.1 | - | 2 - 3 | 247 | 72 | ||
| AJ271671.1 | - | 2 - 3 | 277 | 72 | ||
| AK074943.1 | - | 2 - 3 | 486 | 72 | ||
| AK297983.1 | - | 3 - 4 | 244 | 72 | ||
| AK300244.1 | - | 3 - 4 | 236 | 72 | ||
| BC002563.2 | - | 2 - 3 | 251 | 3138575 | 72 | |
| BC003152.2 | - | 1 - 2 | 223 | 2958651 | 72 | |
| BC007886.2 | - | 2 - 3 | 248 | 4122086 | 72 | |
| BC014303.2 | - | 3 - 4 | 656 | 3942586 | 72 | |
| CR457217.1 | - | 1 - 2 | 190 | 72 | ||
| DA052424.1 | - | 2 - 3 | 370 | 72 | ||
| DA711655.1 | - | 2 - 3 | 379 | 72 | ||
| DA818910.1 | - | 2 - 3 | 356 | 72 | ||
| DB001496.1 | - | 3 - 4 | 430 | 72 | ||
More Information
Set Membership: |
|

SLC39A1
Genomic Map