Assay Details
| Gene Symbol: ANAPC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278486.1 | NP_001265415.1 | 2 - 3 | 213 | 94 | |
| NM_001278487.1 | NP_001265416.1 | 2 - 3 | 213 | 94 | ||
| NM_001278488.1 | NP_001265417.1 | 2 - 3 | 129 | 94 | ||
| NM_001278492.1 | NP_001265421.1 | 2 - 3 | 143 | 94 | ||
| XM_017017491.1 | XP_016872980.1 | 2 - 3 | 226 | 94 | ||
| XM_017017492.1 | XP_016872981.1 | 2 - 3 | 420 | 94 | ||
| XM_017017496.1 | XP_016872985.1 | 2 - 3 | 244 | 94 | ||
| GenBank mRNA | AK222622.1 | - | 2 - 3 | 205 | 94 | |
| BG764016.1 | - | 2 - 3 | 200 | 94 | ||
| BU532056.1 | - | 2 - 3 | 133 | 94 | ||
| BU856100.1 | - | 2 - 3 | 162 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278486.1, NM_001278487.1, NM_001278488.1, NM_001278492.1, XM_017017491.1, XM_017017492.1, XM_017017496.1 transcripts.
Set Membership: |
|

ANAPC15
Genomic Map