Assay Details
| Gene Symbol: ZNF254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278663.1 | NP_001265592.1 | 2 - 3 | 310 | 81 | |
| GenBank mRNA | AK303431.1 | - | 2 - 3 | 244 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK303431.1, NM_001278663.1 transcripts.
Set Membership: |
|

ZNF254
Genomic Map