Assay Details
| Gene Symbol: ZNF254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278665.1 | NP_001265594.1 | 3 - 4 | 397 | 126 | |
| GenBank mRNA | BC056243.1 | - | 3 - 4 | 373 | 126 | |
More Information
Set Membership: |
|

ZNF254
Genomic Map