Assay Details
| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281434.1 | NP_001268363.1 | 2 - 3 | 189 | 86 | |
| GenBank mRNA | BE260293.1 | - | 2 - 3 | 88 | 86 | |
More Information
Set Membership: |
|

ZNHIT3
Genomic Map