Assay Details
| Gene Symbol: VPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282151.1 | NP_001269080.1 | 2 - 3 | 286 | 93 | |
| XM_017019459.1 | XP_016874948.1 | 3 - 4 | 375 | 93 | ||
| GenBank mRNA | CB158821.1 | - | 2 - 3 | 220 | 93 | |
More Information
Set Membership: |
|

VPS29
Genomic Map