Assay Details
| Gene Symbol: ZNF534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143938.1 | NP_001137410.1 | 2 - 3 | 202 | 114 | |
| NM_001291368.1 | NP_001278297.1 | 2 - 3 | 202 | 114 | ||
| GenBank mRNA | AB086839.1 | - | 2 - 3 | 202 | 114 | |
More Information
Set Membership: |
|

ZNF534
Genomic Map