Assay Details
| Gene Symbol: YY1AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198899.1 | NP_001185828.1 | 2 - 3 | 620 | 129 | |
| NM_001198900.1 | NP_001185829.1 | 4 - 5 | 553 | 129 | ||
| NM_001198901.1 | NP_001185830.1 | 4 - 5 | 443 | 129 | ||
| NM_001198902.1 | NP_001185831.1 | 4 - 5 | 410 | 129 | ||
| NM_001198903.1 | NP_001185832.1 | 4 - 5 | 930 | 129 | ||
| NM_001198904.1 | NP_001185833.1 | 4 - 5 | 930 | 129 | ||
| NM_001198905.1 | NP_001185834.1 | 5 - 6 | 594 | 129 | ||
| NM_001198906.1 | NP_001185835.1 | 5 - 6 | 578 | 129 | ||
| NM_018253.3 | NP_060723.2 | 6 - 7 | 675 | 129 | ||
| NM_139118.2 | NP_620829.1 | 5 - 6 | 578 | 129 | ||
| NM_139119.2 | NP_620830.1 | 5 - 6 | 591 | 129 | ||
| NM_139121.2 | NP_620832.1 | 5 - 6 | 544 | 129 | ||
| GenBank mRNA | AF203474.1 | - | 4 - 5 | 610 | 129 | |
| AF206328.1 | - | 5 - 6 | 510 | 129 | ||
| AF466401.1 | - | 4 - 5 | 384 | 129 | ||
| AK001737.1 | - | 1 - 2 | 109 | 129 | ||
| AK023976.1 | - | 5 - 6 | 546 | 129 | ||
| AK225883.1 | - | 1 - 2 | 160 | 129 | ||
| AK297562.1 | - | 4 - 5 | 743 | 129 | ||
| AK298904.1 | - | 5 - 6 | 574 | 129 | ||
| AK303046.1 | - | 4 - 5 | 593 | 129 | ||
| AK303386.1 | - | 2 - 3 | 598 | 129 | ||
| AK309701.1 | - | 4 - 5 | 749 | 129 | ||
| AK315049.1 | - | 5 - 6 | 625 | 129 | ||
| AY604179.1 | - | 4 - 5 | 415 | 129 | ||
| BC001655.2 | - | 6 - 7 | 605 | 2961648 | 129 | |
| BC001843.2 | - | 6 - 7 | 605 | 2961648 | 129 | |
| BC003500.1 | - | 5 - 6 | 544 | 129 | ||
| BC008766.2 | - | 4 - 5 | 487 | 3051391 | 129 | |
| BC009358.2 | - | 1 - 2 | 171 | 129 | ||
| BC014906.2 | - | 5 - 6 | 523 | 3922542 | 129 | |
| BG325066.1 | - | 4 - 5 | 410 | 128 | ||
| CR749789.1 | - | 5 - 6 | 565 | 129 | ||
| DC300265.1 | - | 4 - 5 | 443 | 129 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF206328.1, AK001737.1 transcripts.
Set Membership: |
|

YY1AP1
Genomic Map