Assay Details
| Gene Symbol: ZFAND6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242915.1 | NP_001229844.1 | 1 - 2 | 86 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001242915.1 transcripts.
Set Membership: |
|

ZFAND6
Genomic Map