Assay Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297624.1 | NP_001284553.1 | 1 - 2 | 59 | 85 | |
| NM_001297625.1 | NP_001284554.1 | 1 - 2 | 59 | 85 | ||
| NM_144684.3 | NP_653285.2 | 1 - 2 | 59 | 85 | ||
| GenBank mRNA | AK096442.1 | - | 1 - 2 | 51 | 85 | |
| AK096559.1 | - | 1 - 2 | 51 | 83 | ||
| AK303271.1 | - | 1 - 2 | 50 | 85 | ||
| AL832560.1 | - | 1 - 2 | 38 | 83 | ||
| BC007056.1 | - | 1 - 2 | 56 | 83 | ||
| BC054018.1 | - | 1 - 2 | 59 | 85 | ||
| BC065503.1 | - | 1 - 2 | 36 | 83 | ||
| DA501214.1 | - | 1 - 2 | 59 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK096442.1, AK096559.1, AK303271.1, AL832560.1, BC054018.1, NM_001297624.1, NM_001297625.1, NM_144684.3 transcripts.
Set Membership: |
|

ZNF480
Genomic Map