Assay Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297624.1 | NP_001284553.1 | 3 - 4 | 280 | 129 | |
| GenBank mRNA | AK303271.1 | - | 3 - 4 | 271 | 129 | |
More Information
Set Membership: |
|

ZNF480
Genomic Map