Assay Details
| Gene Symbol: LRP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135772.1 | NP_001129244.1 | 3 - 4 | 787 | 97 | |
| NM_182492.2 | NP_872298.1 | 1 - 2 | 2728 | 97 | ||
| XM_006724372.3 | XP_006724435.1 | 3 - 4 | 1245 | 97 | ||
| XM_017029083.1 | XP_016884572.1 | 4 - 5 | 3165 | 97 | ||
| XM_017029084.1 | XP_016884573.1 | 3 - 4 | 3389 | 97 | ||
| XM_017029085.1 | XP_016884574.1 | 4 - 5 | 997 | 97 | ||
| XM_017029086.1 | XP_016884575.1 | 4 - 5 | 3316 | 97 | ||
| XM_017029087.1 | XP_016884576.1 | 2 - 3 | 5940 | 97 | ||
| XM_017029088.1 | XP_016884577.1 | 3 - 4 | 3413 | 97 | ||
| GenBank mRNA | AL137651.1 | - | 1 - 2 | 2730 | 97 | |
| BC091519.1 | - | 1 - 2 | 986 | 97 | ||
| BC137460.1 | - | 1 - 2 | 2102 | 9021460 | 97 | |
| BC137463.1 | - | 1 - 2 | 2102 | 9021463 | 97 | |
| BC139736.1 | - | 3 - 4 | 793 | 40034226 | 97 | |
More Information
Set Membership: |
|

LRP5L
Genomic Map