Assay Details
| Gene Symbol: ZNF773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304334.1 | NP_001291263.1 | 3 - 4 | 490 | 100 | |
| NM_198542.2 | NP_940944.1 | 3 - 4 | 493 | 100 | ||
| GenBank mRNA | AK057209.1 | - | 3 - 4 | 409 | 100 | |
More Information
Set Membership: |
|

ZNF773
Genomic Map