Assay Details
| Gene Symbol: DPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206831.2 | NP_996662.1 | 2 - 3 | 292 | 85 | |
| GenBank mRNA | AK092040.1 | - | 2 - 3 | 245 | 85 | |
| AK311618.1 | - | 2 - 3 | 254 | 85 | ||
| BC010181.2 | - | 2 - 3 | 201 | 4543414 | 85 | |
| DA875039.1 | - | 2 - 3 | 292 | 85 | ||
More Information
Set Membership: |
|

DPH3
Genomic Map