Assay Details
| Gene Symbol: ATAD3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170535.1 | NP_001164006.1 | 8 | 920 | 96 | |
| NM_001170536.1 | NP_001164007.1 | 8 | 750 | 96 | ||
| NM_018188.3 | NP_060658.3 | 8 | 1064 | 96 | ||
| XM_017001628.1 | XP_016857117.1 | 8 | 920 | 96 | ||
| GenBank mRNA | AK001571.1 | - | 8 | 903 | 96 | |
| AK056099.1 | - | 8 | 750 | 96 | ||
| AK091144.1 | - | 8 | 1032 | 96 | ||
| AK310572.1 | - | 942 | 96 | |||
| BC007803.2 | - | 8 | 881 | 4136170 | 96 | |
| BC011814.2 | - | 8 | 886 | 3162989 | 96 | |
| BC014101.2 | - | 8 | 910 | 3503438 | 96 | |
| BC033109.1 | - | 8 | 884 | 4877449 | 96 | |
| BC063607.1 | - | 8 | 878 | 4939339 | 96 | |
| GU189416.1 | - | 8 | 824 | 96 | ||
| Gene Symbol: ATAD3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF343078.1 | - | 8 | 857 | 96 | |
More Information
Set Membership: |
|

ATAD3A
Genomic Map