Assay Details
| Gene Symbol: ZNF254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278661.1 | NP_001265590.1 | 5 | 4084 | 141 | |
| NM_001278662.1 | NP_001265591.1 | 5 | 4064 | 141 | ||
| NM_001278663.1 | NP_001265592.1 | 3 | 3861 | 141 | ||
| NM_001278664.1 | NP_001265593.1 | 3 | 3822 | 141 | ||
| NM_001278677.1 | NP_001265606.1 | 6 | 4156 | 141 | ||
| NM_001278678.1 | NP_001265607.1 | 2 | 3726 | 141 | ||
| NM_203282.3 | NP_975011.3 | 4 | 3949 | 141 | ||
| GenBank mRNA | BM976089.1 | - | 1 | 179 | 141 | |
More Information
Set Membership: |
|

ZNF254
Genomic Map