Assay Details
| Gene Symbol: ZNF254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278661.1 | NP_001265590.1 | 5 | 3077 | 82 | |
| NM_001278662.1 | NP_001265591.1 | 5 | 3057 | 82 | ||
| NM_001278663.1 | NP_001265592.1 | 3 | 2854 | 82 | ||
| NM_001278664.1 | NP_001265593.1 | 3 | 2815 | 82 | ||
| NM_001278677.1 | NP_001265606.1 | 6 | 3149 | 82 | ||
| NM_001278678.1 | NP_001265607.1 | 2 | 2719 | 82 | ||
| NM_203282.3 | NP_975011.3 | 4 | 2942 | 82 | ||
| XM_011528443.1 | XP_011526745.1 | 6 | 3210 | 82 | ||
| XM_011528444.1 | XP_011526746.1 | 7 | 3230 | 82 | ||
| XM_011528448.2 | XP_011526750.1 | 4 | 2889 | 82 | ||
| XM_017027514.1 | XP_016883003.1 | 6 | 3140 | 82 | ||
| XM_017027515.1 | XP_016883004.1 | 8 | 3567 | 82 | ||
| XM_017027516.1 | XP_016883005.1 | 4 | 5614 | 82 | ||
| XM_017027517.1 | XP_016883006.1 | 5 | 3443 | 82 | ||
| XM_017027518.1 | XP_016883007.1 | 4 | 2939 | 82 | ||
| XM_017027519.1 | XP_016883008.1 | 5 | 3218 | 82 | ||
| GenBank mRNA | AK303431.1 | - | 3 | 2788 | 82 | |
| AL832713.1 | - | 1 | 1638 | 82 | ||
| BC043147.1 | - | 4 | 2914 | 5296940 | 82 | |
| BU783464.1 | - | 1 | 115 | 82 | ||
| CN395850.1 | - | 1 | 433 | 82 | ||
More Information
Set Membership: |
|

ZNF254
Genomic Map