Assay Details
| Gene Symbol: TUBB4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006088.5 | NP_006079.1 | 4 | 1125 | 84 | |
| GenBank mRNA | AK000560.1 | - | 4 | 1097 | 84 | |
| AK021414.1 | - | 4 | 953 | 84 | ||
| AK026167.1 | - | 4 | 1096 | 84 | ||
| AK026594.1 | - | 4 | 1097 | 84 | ||
| AK026609.1 | - | 4 | 1097 | 84 | ||
| AK098686.1 | - | 4 | 1026 | 84 | ||
| AY167990.1 | - | 4 | 1023 | 84 | ||
| BC001911.1 | - | 4 | 1060 | 3544236 | 84 | |
| BC002783.1 | - | 4 | 1070 | 3614023 | 84 | |
| BC002885.2 | - | 4 | 1067 | 3944735 | 84 | |
| BC004188.2 | - | 4 | 1072 | 2964559 | 84 | |
| BC007889.2 | - | 4 | 1061 | 4127195 | 84 | |
| BC008006.1 | - | 1 | 371 | 84 | ||
| BC012835.2 | - | 4 | 1072 | 3603755 | 84 | |
| BC019359.1 | - | 4 | 1063 | 4024979 | 84 | |
| BC019829.1 | - | 4 | 1086 | 5110038 | 84 | |
| BC024038.1 | - | 4 | 1061 | 3616531 | 84 | |
| BC029529.1 | - | 4 | 1097 | 5274343 | 84 | |
| BC039175.1 | - | 4 | 1070 | 4385885 | 84 | |
| BC071889.1 | - | 4 | 1061 | 4905049 | 84 | |
| BX648521.1 | - | 3 | 1571 | 84 | ||
More Information
Set Membership: |
|

TUBB4B
Genomic Map