Assay Details
| Gene Symbol: MAP1LC3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022818.4 | NP_073729.1 | 4 | 1523 | 148 | |
| GenBank mRNA | AF087871.1 | - | 4 | 1382 | 148 | |
| AF303888.1 | - | 4 | 1328 | 148 | ||
| AK024571.1 | - | 1 | 660 | 148 | ||
| AK025556.1 | - | 4 | 1374 | 148 | ||
| AL834298.1 | - | 4 | 1292 | 148 | ||
| BC018634.2 | - | 4 | 1361 | 3623259 | 148 | |
| BC041874.1 | - | 4 | 1918 | 5271912 | 148 | |
| BC045759.1 | - | 5 | 1864 | 4828857 | 148 | |
| BC048009.1 | - | 4 | 1407 | 148 | ||
| BC067797.1 | - | 4 | 1373 | 5276841 | 148 | |
More Information
Set Membership: |
|

MAP1LC3B
Genomic Map