Assay Details
| Gene Symbol: UGT2B17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077.3 | NP_001068.1 | 6 | 1817 | 199 | |
| GenBank mRNA | U59209.1 | - | 6 | 1825 | 199 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT2B17| Gene Symbol: UGT2B17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077.3 | NP_001068.1 | 6 | 1817 | 199 | |
| GenBank mRNA | U59209.1 | - | 6 | 1825 | 199 | |
Set Membership: |
|