Assay Details
| Gene Symbol: SH3BP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046084.1 | 1 - 2 | 3422 | 114 | ||
| GenBank mRNA | AL133111.1 | - | 1 - 2 | 3423 | 114 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calpain 7 | CAPN7 |
CALPAIN7, PALBH |
-1.3kb | Sense | Chr. 3 - 15206226 - 15252916 on NCBI Build GRCh38 |
| Downstream | methyltransferase like 6 | METTL6 | 120.7kb | Anti-sense | Chr. 3 - 15381275 - 15427580 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SH3BP5-AS1
Genomic Map