Assay Details
| Gene Symbol: CCL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005623.2 | NP_005614.2 | 1 - 2 | 533 | 72 | |
| GenBank mRNA | BC126242.1 | - | 1 - 2 | 340 | 8991958 | 72 |
| Y10802.1 | - | 1 - 2 | 134 | 72 | ||
| Y16645.1 | - | 1 - 2 | 550 | 72 | ||
More Information
Set Membership: |
|

CCL8
Genomic Map