Assay Details
| Gene Symbol: IL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000589.3 | NP_000580.1 | 3 - 4 | 427 | 141 | |
| NM_172348.2 | NP_758858.1 | 2 - 3 | 379 | 141 | ||
| GenBank mRNA | AB102862.1 | - | 2 - 3 | 314 | 141 | |
| AM937235.1 | - | 3 - 4 | 402 | 141 | ||
| BC066278.1 | - | 3 - 4 | 427 | 141 | ||
| BC067514.1 | - | 3 - 4 | 427 | 6971780 | 141 | |
| BC067515.1 | - | 3 - 4 | 427 | 6971782 | 141 | |
| BC070123.1 | - | 3 - 4 | 424 | 30415895 | 141 | |
| CD640452.1 | - | 3 - 4 | 427 | 141 | ||
| JX878389.1 | - | 4 - 5 | 463 | 141 | ||
| M13982.1 | - | 3 - 4 | 425 | 141 | ||
| X81851.1 | - | 2 - 3 | 314 | 141 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

IL4
Genomic Map