Assay Details
| Gene Symbol: IL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000879.2 | NP_000870.1 | 2 - 3 | 217 | 95 | |
| XM_005271988.3 | XP_005272045.1 | 3 - 4 | 351 | 95 | ||
| XM_011543373.2 | XP_011541675.1 | 4 - 5 | 453 | 95 | ||
| XM_011543374.2 | XP_011541676.1 | 3 - 4 | 404 | 95 | ||
| GenBank mRNA | BC066282.1 | - | 2 - 3 | 196 | 6971771 | 95 |
| X04688.1 | - | 2 - 3 | 217 | 95 | ||
| X12705.1 | - | 2 - 3 | 217 | 95 | ||
More Information
Set Membership: |
|

IL5
Genomic Map