Assay Details
| Gene Symbol: IL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000882.3 | NP_000873.2 | 2 | 448 | 144 | |
| GenBank mRNA | AF101062.1 | - | 2 | 227 | 144 | |
| AF180562.1 | - | 2 | 286 | 144 | ||
| BC104982.1 | - | 2 | 380 | 8143985 | 144 | |
| BC104984.1 | - | 2 | 380 | 8143987 | 144 | |
| M65271.1 | - | 2 | 294 | 144 | ||
| M65291.1 | - | 2 | 328 | 144 | ||
More Information
Set Membership: |
|

IL12A
Genomic Map