Assay Details
| Gene Symbol: IL12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002187.2 | NP_002178.2 | 3 | 374 | 120 | |
| GenBank mRNA | AF180563.1 | - | 2 | 338 | 120 | |
| BC067498.1 | - | 2 | 336 | 6971868 | 120 | |
| BC067499.1 | - | 2 | 336 | 6971869 | 120 | |
| BC067500.1 | - | 2 | 336 | 6971872 | 120 | |
| BC067501.1 | - | 2 | 336 | 6971873 | 120 | |
| BC067502.1 | - | 2 | 336 | 6971874 | 120 | |
| BC074723.2 | - | 3 | 363 | 30915173 | 120 | |
| M65272.1 | - | 3 | 374 | 120 | ||
| M65290.1 | - | 2 | 345 | 120 | ||
More Information
Set Membership: |
|

IL12B
Genomic Map