Assay Details
| Gene Symbol: LTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002341.1 | NP_002332.1 | 3 | 252 | 146 | |
| GenBank mRNA | BC069330.1 | - | 3 | 252 | 7262211 | 146 |
| L11015.1 | - | 3 | 252 | 146 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001530.hg.1, TC6_apd_hap1000081.hg.1, TC6_cox_hap2000159.hg.1, TC6_dbb_hap3000148.hg.1, TC6_mann_hap4000136.hg.1, TC6_mcf_hap5000136.hg.1, TC6_qbl_hap6000151.hg.1, TC6_ssto_hap7000131.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
immune response
signal transduction
cell-cell signaling
gene expression
tumor necrosis factor-mediated signaling pathway
skin development
positive regulation of interleukin-12 biosynthetic process
lymph node development
receptor binding
cytokine activity
tumor necrosis factor receptor binding
immune response
signal transduction
cell-cell signaling
gene expression
tumor necrosis factor-mediated signaling pathway
skin development
positive regulation of interleukin-12 biosynthetic process
lymph node development
receptor binding
cytokine activity
tumor necrosis factor receptor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06017505.hg.1 | TC06001530.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017505.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017505.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017505.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017505.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06017505.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002749.hg.1 | TC6_dbb_hap3000148.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002749.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3002749.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002365.hg.1 | TC6_mann_hap4000136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002365.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4002365.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002506.hg.1 | TC6_mcf_hap5000136.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002506.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5002506.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001154.hg.1 | TC6_apd_hap1000081.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001154.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1001154.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002835.hg.1 | TC6_cox_hap2000159.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002835.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2002835.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002817.hg.1 | TC6_qbl_hap6000151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002817.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6002817.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002505.hg.1 | TC6_ssto_hap7000131.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002505.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7002505.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011438.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | ENST00000429299.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | ENST00000446745.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | ENST00000482429.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | ENST00000483972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | NM_002341.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | NM_009588.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600177277.hg.1 | TC0600011438.hg.1 | Clariom D TC |

LTB
Genomic Map