Assay Details
| Gene Symbol: HSPA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005347.4 | NP_005338.1 | 1 - 2 | 383 | 101 | |
| GenBank mRNA | AF216292.1 | - | 1 - 2 | 122 | 101 | |
| AJ271729.1 | - | 1 - 2 | 135 | 101 | ||
| AK293604.1 | - | 1 - 2 | 339 | 101 | ||
| BC020235.1 | - | 1 - 2 | 326 | 5020098 | 101 | |
| DA549454.1 | - | 1 - 2 | 489 | 101 | ||
| EU794617.1 | - | 1 - 2 | 135 | 101 | ||
| X87949.1 | - | 1 - 2 | 344 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001589.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Citations (2)
Protein expression profiling in esophageal adenocarcinoma patients indicates association of heat-shock protein 27 expression and chemotherapy response.
- Authors
- Langer R, Ott K, Specht K, Becker K, Lordick F, Burian M, Herrmann K, Schrattenholz A, Cahill MA, Schwaiger M, Hofler H, Wester HJ
- Journal
- Clin Cancer Res - Page No 8279 (Mon Dec 15 00:00:00 EST 2008)
- Volume (Year)
- 14 (2008)
- Product Usage
- -
Expression and clinical significance of glucose regulated proteins GRP78 (BiP) and GRP94 (GP96) in human adenocarcinomas of the esophagus.
- Authors
- Langer R, Feith M, Siewert JR, Wester HJ, Hoefler H
- Journal
- Bmc Cancer - Page No 70 (Tue Jan 01 00:00:00 EST 2008)
- Volume (Year)
- 8 (2008)
- Product Usage
- -

HSPA5
Genomic Map