Assay Details
| Gene Symbol: TBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK298272.1 | - | 7 | 904 | 127 | |
| AK304648.1 | - | 5 | 807 | 127 | ||
| BC109053.1 | - | 6 | 933 | 40032492 | 127 | |
| BT019657.1 | - | 5 | 734 | 127 | ||
| M34960.1 | - | 5 | 743 | 127 | ||
| M55654.1 | - | 6 | 984 | 127 | ||
| X54993.1 | - | 5 | 829 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Citations (27)
Disturbed expression of phase I and phase II estrogen-metabolizing enzymes in endometrial cancer: lower levels of CYP1B1 and increased expression of S-COMT.
- Authors
- Hevir N, Sinkovec J, Rizner TL
- Journal
- Mol Cell Endocrinol - Page No 158 (Sat Jan 01 00:00:00 EST 2011)
- Volume (Year)
- 331 (2011)
- Product Usage
- -
Loss of p75 neurotrophin receptor expression accompanies malignant progression to human and murine retinoblastoma.
- Authors
- Dimaras H, Coburn B, Pajovic S, Gallie BL
- Journal
- Mol Carcinog - Page No 333 (Mon May 01 00:00:00 EDT 2006)
- Volume (Year)
- 45 (2006)
- Product Usage
- -
Anagrelide represses GATA-1 and FOG-1 expression without interfering with thrombopoietin receptor signal transduction.
- Authors
- Ahluwalia M, Donovan H, Singh N, Butcher L, Erusalimsky JD
- Journal
- J Thromb Haemost - Page No 2252 (Fri Oct 01 00:00:00 EDT 2010)
- Volume (Year)
- 8 (2010)
- Product Usage
- -
Mechanisms of gene amplification and evidence of coamplification in drug-resistant human osteosarcoma cell lines.
- Authors
- Hattinger CM, Stoico G, Michelacci F, Pasello M, Scionti I, Remondini D, Castellani GC, Fanelli M, Scotlandi K, Picci P, Serra M
- Journal
- Genes Chromosomes Cancer - Page No 289 (Wed Apr 01 00:00:00 EDT 2009)
- Volume (Year)
- 48 (2009)
- Product Usage
- -
Acute in vivo effects of insulin on gene expression in adipose tissue in insulin-resistant and insulin-sensitive subjects.
- Authors
- Westerbacka J, Corner A, Kannisto K, Kolak M, Makkonen J, Korsheninnikova E, Nyman T, Hamsten A, Fisher RM, Yki-Jarvinen H
- Journal
- Diabetologia - Page No 132 (Sun Jan 01 00:00:00 EST 2006)
- Volume (Year)
- 49 (2006)
- Product Usage
- -
Modulation of Na+-Ca2+ exchanger expression by immunosuppressive drugs is isoform-specific.
- Authors
- Elbaz B, Alperovitch A, Gottesman MM, Kimchi-Sarfaty C, Rahamimoff H
- Journal
- Mol Pharmacol - Page No 1254 (Tue Apr 01 00:00:00 EDT 2008)
- Volume (Year)
- 73 (2008)
- Product Usage
- -
KIF14 messenger RNA expression is independently prognostic for outcome in lung cancer.
- Authors
- Corson TW, Zhu CQ, Lau SK, Shepherd FA, Tsao MS, Gallie BL
- Journal
- Clin Cancer Res - Page No 3229 (Fri Jun 01 00:00:00 EDT 2007)
- Volume (Year)
- 13 (2007)
- Product Usage
- -
AMPA receptor subunit and splice variant expression in the DLPFC of schizophrenic subjects and rhesus monkeys chronically administered antipsychotic drugs.
- Authors
- O'Connor JA, Muly EC, Arnold SE, Hemby SE
- Journal
- Schizophr Res - Page No 28 (Thu Feb 01 00:00:00 EST 2007)
- Volume (Year)
- 90 (2007)
- Product Usage
- -
Parkinson patient fibroblasts show increased alpha-synuclein expression.
- Authors
- Hoepken HH, Gispert S, Azizov M, Klinkenberg M, Ricciardi F, Kurz A, Morales-Gordo B, Bonin M, Riess O, Gasser T, Kogel D, Steinmetz H, Auburger G
- Journal
- Exp Neurol - Page No 307 (Fri Aug 01 00:00:00 EDT 2008)
- Volume (Year)
- 212 (2008)
- Product Usage
- -
Prolactin and oestrogen synergistically regulate gene expression and proliferation of breast cancer cells.
- Authors
- Rasmussen LM, Frederiksen KS, Din N, Galsgaard E, Christensen L, Berchtold MW, Panina S
- Journal
- Endocr Relat Cancer - Page No 809 (Wed Sep 01 00:00:00 EDT 2010)
- Volume (Year)
- 17 (2010)
- Product Usage
- -
Elevated GRIA1 mRNA expression in Layer II/III and V pyramidal cells of the DLPFC in schizophrenia.
- Authors
- O'Connor JA, Hemby SE
- Journal
- Schizophr Res - Page No 277 (Sat Dec 01 00:00:00 EST 2007)
- Volume (Year)
- 97 (2007)
- Product Usage
- -
Cytoglobin is upregulated by tumour hypoxia and silenced by promoter hypermethylation in head and neck cancer.
- Authors
- Shaw RJ, Omar MM, Rokadiya S, Kogera FA, Lowe D, Hall GL, Woolgar JA, Homer J, Liloglou T, Field JK, Risk JM
- Journal
- Br J Cancer - Page No 139 (Tue Jul 07 00:00:00 EDT 2009)
- Volume (Year)
- 101 (2009)
- Product Usage
- -
Effects of chronic rosiglitazone therapy on gene expression in human adipose tissue in vivo in patients with type 2 diabetes.
- Authors
- Kolak M, Yki-Jarvinen H, Kannisto K, Tiikkainen M, Hamsten A, Eriksson P, Fisher RM
- Journal
- J Clin Endocrinol Metab - Page No 720 (Thu Feb 01 00:00:00 EST 2007)
- Volume (Year)
- 92 (2007)
- Product Usage
- -
Mitochondrial dysfunction, peroxidation damage and changes in glutathione metabolism in PARK6.
- Authors
- Hoepken HH, Gispert S, Morales B, Wingerter O, Del Turco D, Mulsch A, Nussbaum RL, Muller K, Drose S, Brandt U, Deller T, Wirth B, Kudin AP, Kunz WS, Auburger G
- Journal
- Neurobiol Dis - Page No 401 (Thu Feb 01 00:00:00 EST 2007)
- Volume (Year)
- 25 (2007)
- Product Usage
- -
Gene expression profile for predicting survival in advanced-stage serous ovarian cancer across two independent datasets.
- Authors
- Yoshihara K, Tajima A, Yahata T, Kodama S, Fujiwara H, Suzuki M, Onishi Y, Hatae M, Sueyoshi K, Fujiwara H, Kudo Y, Kotera K, Masuzaki H, Tashiro H, Katabuchi H, Inoue I, Tanaka K
- Journal
- Plos One - Page No e9615 (Fri Jan 01 00:00:00 EST 2010)
- Volume (Year)
- 5 (2010)
- Product Usage
- -
Evaluating reference genes to normalize gene expression in human epileptogenic brain tissues.
- Authors
- Wierschke S, Gigout S, Horn P, Lehmann TN, Dehnicke C, Brauer AU, Deisz RA
- Journal
- Biochem Biophys Res Commun - Page No (Fri Nov 19 00:00:00 EST 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
Novel porcine housekeeping genes for real-time RT-PCR experiments normalization in adipose tissue: Assessment of leptin mRNA quantity in different pig breeds.
- Authors
- Piorkowska K, Oczkowicz M, Rozycki M, Ropka-Molik K, Kajtoch AP
- Journal
- Meat Sci - Page No (Sat Oct 16 00:00:00 EDT 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
High expression of KIF14 in retinoblastoma: association with older age at diagnosis.
- Authors
- Madhavan J, Coral K, Mallikarjuna K, Corson TW, Amit N, Khetan V, George R, Biswas J, Gallie BL, Kumaramanickavel G
- Journal
- Invest Ophthalmol Vis Sci - Page No 4901 (Thu Nov 01 00:00:00 EDT 2007)
- Volume (Year)
- 48 (2007)
- Product Usage
- -
Microarray analysis of gene expression in peripheral blood mononuclear cells from dioxin-exposed human subjects.
- Authors
- McHale CM, Zhang L, Hubbard AE, Zhao X, Baccarelli A, Pesatori AC, Smith MT, Landi MT
- Journal
- Toxicology - Page No 101 (Fri Jan 05 00:00:00 EST 2007)
- Volume (Year)
- 229 (2007)
- Product Usage
- -
The hypoxia-inducible factor 1/NOR-1 axis regulates the survival response of endothelial cells to hypoxia.
- Authors
- Martorell L, Gentile M, Rius J, Rodriguez C, Crespo J, Badimon L, Martinez-Gonzalez J
- Journal
- Mol Cell Biol - Page No 5828 (Sun Nov 01 00:00:00 EDT 2009)
- Volume (Year)
- 29 (2009)
- Product Usage
- -
Circulating and placental growth-differentiation factor 15 in preeclampsia and in pregnancy complicated by diabetes mellitus.
- Authors
- Sugulle M, Dechend R, Herse F, Weedon-Fekjaer MS, Johnsen GM, Brosnihan KB, Anton L, Luft FC, Wollert KC, Kempf T, Staff AC
- Journal
- Hypertension - Page No 106 (Wed Jul 01 00:00:00 EDT 2009)
- Volume (Year)
- 54 (2009)
- Product Usage
- -
Enhanced vulnerability of PARK6 patient skin fibroblasts to apoptosis induced by proteasomal stress.
- Authors
- Klinkenberg M, Thurow N, Gispert S, Ricciardi F, Eich F, Prehn JH, Auburger G, Kogel D
- Journal
- Neuroscience - Page No 422 (Wed Mar 17 00:00:00 EDT 2010)
- Volume (Year)
- 166 (2010)
- Product Usage
- -
Reduced in vitro functional activity of human NRAMP1 (SLC11A1) allele that predisposes to increased risk of pediatric tuberculosis disease.
- Authors
- Gallant CJ, Malik S, Jabado N, Cellier M, Simkin L, Finlay BB, Graviss EA, Gros P, Musser JM, Schurr E
- Journal
- Genes Immun - Page No 691 (Sat Dec 01 00:00:00 EST 2007)
- Volume (Year)
- 8 (2007)
- Product Usage
- -
Gene expression by human monocytes from peripheral blood in response to exposure to metals.
- Authors
- Jost-Albrecht K, Hofstetter W
- Journal
- J Biomed Mater Res B Appl Biomater - Page No 449 (Wed Feb 01 00:00:00 EST 2006)
- Volume (Year)
- 76 (2006)
- Product Usage
- -
Distinction in gene expression profiles demonstrated in parathyroid adenomas by high-density oligoarray technology.
- Authors
- Forsberg L, Bjorck E, Hashemi J, Zedenius J, Hoog A, Farnebo LO, Reimers M, Larsson C
- Journal
- Eur J Endocrinol - Page No 459 (Tue Mar 01 00:00:00 EST 2005)
- Volume (Year)
- 152 (2005)
- Product Usage
- -
Lysyl oxidase activity in the ocular tissues and the role of LOX in proliferative diabetic retinopathy and rhegmatogenous retinal detachment.
- Authors
- Coral K, Angayarkanni N, Madhavan J, Bharathselvi M, Ramakrishnan S, Nandi K, Rishi P, Kasinathan N, Krishnakumar S
- Journal
- Invest Ophthalmol Vis Sci - Page No 4746 (Sat Nov 01 00:00:00 EDT 2008)
- Volume (Year)
- 49 (2008)
- Product Usage
- -
Inhibition of telomerase in the endothelial cells disrupts tumor angiogenesis in glioblastoma xenografts.
- Authors
- Falchetti ML, Mongiardi MP, Fiorenzo P, Petrucci G, Pierconti F, D'Agnano I, D'Alessandris G, Alessandri G, Gelati M, Ricci-Vitiani L, Maira G, Larocca LM, Levi A, Pallini R
- Journal
- Int J Cancer - Page No 1236 (Sat Mar 15 00:00:00 EDT 2008)
- Volume (Year)
- 122 (2008)
- Product Usage
- -

TBP
Genomic Map