Assay Details
| Gene Symbol: TGFB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000660.5 | NP_000651.3 | 4 - 5 | 1604 | 125 | |
| GenBank mRNA | AK291907.1 | - | 4 - 5 | 1093 | 125 | |
| AK307742.1 | - | 4 - 5 | 1289 | 125 | ||
| BC000125.1 | - | 4 - 5 | 1161 | 3351664 | 125 | |
| BC001180.1 | - | 4 - 5 | 1161 | 3356605 | 125 | |
| BC022242.1 | - | 4 - 5 | 1084 | 4399762 | 125 | |
| BT007245.1 | - | 4 - 5 | 715 | 125 | ||
| X02812.1 | - | 4 - 5 | 1559 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (19)
Systemic and local interleukin-17 and linked cytokines associated with Sjogren's syndrome immunopathogenesis.
- Authors
- Katsifis GE, Rekka S, Moutsopoulos NM, Pillemer S, Wahl SM
- Journal
- Am J Pathol - Page No 1167 (Tue Sep 01 00:00:00 EDT 2009)
- Volume (Year)
- 175 (2009)
- Product Usage
- -
Asporin and transforming growth factor-beta gene expression in osteoblasts from subchondral bone and osteophytes in osteoarthritis.
- Authors
- Sakao K, Takahashi KA, Arai Y, Saito M, Honjyo K, Hiraoka N, Kishida T, Mazda O, Imanishi J, Kubo T
- Journal
- J Orthop Sci - Page No 738 (Sun Nov 01 00:00:00 EDT 2009)
- Volume (Year)
- 14 (2009)
- Product Usage
- -
The synthetic liver X receptor agonist GW3965 reduces tissue factor production and inflammatory responses in human islets in vitro.
- Authors
- Scholz H, Lund T, Dahle MK, Collins JL, Korsgren O, Wang JE, Foss A
- Journal
- Diabetologia - Page No 1352 (Wed Jul 01 00:00:00 EDT 2009)
- Volume (Year)
- 52 (2009)
- Product Usage
- -
Role for staphylococci in misguided thrombus resolution of chronic thromboembolic pulmonary hypertension.
- Authors
- Bonderman D, Jakowitsch J, Redwan B, Bergmeister H, Renner MK, Panzenbock H, Adlbrecht C, Georgopoulos A, Klepetko W, Kneussl M, Lang IM
- Journal
- Arterioscler Thromb Vasc Biol - Page No 678 (Tue Apr 01 00:00:00 EDT 2008)
- Volume (Year)
- 28 (2008)
- Product Usage
- -
Pattern of expression of inflammatory markers in adipose tissue of untreated hypertensive patients.
- Authors
- Madec S, Chiarugi M, Santini E, Rossi C, Miccoli P, Ferrannini E, Solini A
- Journal
- J Hypertens - Page No 1459 (Thu Jul 01 00:00:00 EDT 2010)
- Volume (Year)
- 28 (2010)
- Product Usage
- -
Alterations in intestinal fatty acid metabolism in inflammatory bowel disease.
- Authors
- Heimerl S, Moehle C, Zahn A, Boettcher A, Stremmel W, Langmann T, Schmitz G
- Journal
- Biochim Biophys Acta - Page No 341 (Wed Mar 01 00:00:00 EST 2006)
- Volume (Year)
- 1762 (2006)
- Product Usage
- -
Hyaluronic acid modulates gene expression of connective tissue growth factor (CTGF), transforming growth factor-beta1 (TGF-beta1), and vascular endothelial growth factor (VEGF) in human fibroblast-like synovial cells from advanced-stage osteoarthritis in vitro.
- Authors
- Lee YT, Shao HJ, Wang JH, Liu HC, Hou SM, Young TH
- Journal
- J Orthop Res - Page No 492 (Thu Apr 01 00:00:00 EDT 2010)
- Volume (Year)
- 28 (2010)
- Product Usage
- -
Cyclic mechanical stretch reduces myofibroblast differentiation of primary lung fibroblasts.
- Authors
- Blaauboer ME, Smit TH, Hanemaaijer R, Stoop R, Everts V
- Journal
- Biochem Biophys Res Commun - Page No (Sat Nov 20 00:00:00 EST 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
Induction of Foxp3+ regulatory T cells with histone deacetylase inhibitors.
- Authors
- Lucas JL, Mirshahpanah P, Haas-Stapleton E, Asadullah K, Zollner TM, Numerof RP
- Journal
- Cell Immunol - Page No 97 (Thu Jan 01 00:00:00 EST 2009)
- Volume (Year)
- 257 (2009)
- Product Usage
- -
Differential expression patterns of cytokines in complex regional pain syndrome.
- Authors
- Uceyler N, Eberle T, Rolke R, Birklein F, Sommer C
- Journal
- Pain - Page No 195 (Thu Nov 01 00:00:00 EDT 2007)
- Volume (Year)
- 132 (2007)
- Product Usage
- -
The phenotypic responses of human anterior cruciate ligament cells cultured on poly(epsilon-caprolactone) and chitosan.
- Authors
- Shao HJ, Chen CS, Lee YT, Wang JH, Young TH
- Journal
- J Biomed Mater Res A - Page No 1297 (Tue Jun 15 00:00:00 EDT 2010)
- Volume (Year)
- 93 (2010)
- Product Usage
- -
Paucity of pericytes in germinal matrix vasculature of premature infants.
- Authors
- Braun A, Xu H, Hu F, Kocherlakota P, Siegel D, Chander P, Ungvari Z, Csiszar A, Nedergaard M, Ballabh P
- Journal
- J Neurosci - Page No 12012 (Wed Oct 31 00:00:00 EDT 2007)
- Volume (Year)
- 27 (2007)
- Product Usage
- -
Extracorporeal photochemotherapy is accompanied by increasing levels of circulating CD4+CD25+GITR+Foxp3+CD62L+ functional regulatory T-cells in patients with graft-versus-host disease.
- Authors
- Biagi E, Di Biaso I, Leoni V, Gaipa G, Rossi V, Bugarin C, Renoldi G, Parma M, Balduzzi A, Perseghin P, Biondi A
- Journal
- Transplantation - Page No 31 (Sun Jul 15 00:00:00 EDT 2007)
- Volume (Year)
- 84 (2007)
- Product Usage
- -
The antifibrotic drug halofuginone inhibits proliferation and collagen production by human leiomyoma and myometrial smooth muscle cells.
- Authors
- Grudzien MM, Low PS, Manning PC, Arredondo M, Belton RJ Jr, Nowak RA
- Journal
- Fertil Steril - Page No 1290 (Mon Mar 01 00:00:00 EST 2010)
- Volume (Year)
- 93 (2010)
- Product Usage
- -
Reversing the defective induction of IL-10-secreting regulatory T cells in glucocorticoid-resistant asthma patients.
- Authors
- Xystrakis E, Kusumakar S, Boswell S, Peek E, Urry Z, Richards DF, Adikibi T, Pridgeon C, Dallman M, Loke TK, Robinson DS, Barrat FJ, O'Garra A, Lavender P, Lee TH, Corrigan C, Hawrylowicz CM
- Journal
- J Clin Invest - Page No 146 (Sun Jan 01 00:00:00 EST 2006)
- Volume (Year)
- 116 (2006)
- Product Usage
- -
Mesenchymal stem cell ingrowth and differentiation on coralline hydroxyapatite scaffolds.
- Authors
- Mygind T, Stiehler M, Baatrup A, Li H, Zou X, Flyvbjerg A, Kassem M, Bunger C
- Journal
- Biomaterials - Page No 1036 (Thu Feb 01 00:00:00 EST 2007)
- Volume (Year)
- 28 (2007)
- Product Usage
- -
Modulation of gene expression and collagen production of anterior cruciate ligament cells through cell shape changes on polycaprolactone/chitosan blends.
- Authors
- Shao HJ, Lee YT, Chen CS, Wang JH, Young TH
- Journal
- Biomaterials - Page No 4695 (Tue Jun 01 00:00:00 EDT 2010)
- Volume (Year)
- 31 (2010)
- Product Usage
- -
Myeloid dendritic cells from human cutaneous squamous cell carcinoma are poor stimulators of T-cell proliferation.
- Authors
- Bluth MJ, Zaba LC, Moussai D, Suarez-Farinas M, Kaporis H, Fan L, Pierson KC, White TR, Pitts-Kiefer A, Fuentes-Duculan J, Guttman-Yassky E, Krueger JG, Lowes MA, Carucci JA
- Journal
- J Invest Dermatol - Page No 2451 (Thu Oct 01 00:00:00 EDT 2009)
- Volume (Year)
- 129 (2009)
- Product Usage
- -
Effect of dynamic 3-D culture on proliferation, distribution, and osteogenic differentiation of human mesenchymal stem cells.
- Authors
- Stiehler M, Bunger C, Baatrup A, Lind M, Kassem M, Mygind T
- Journal
- J Biomed Mater Res A - Page No 96 (Wed Apr 01 00:00:00 EDT 2009)
- Volume (Year)
- 89 (2009)
- Product Usage
- -

TGFB1
Genomic Map