Assay Details
| Gene Symbol: PPP2CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285132.1 | NP_001272061.1 | 3 - 4 | 699 | 60 | |
| XM_015451350.1 | XP_015306836.1 | 3 - 4 | 492 | 60 | ||
| GenBank mRNA | AB169507.1 | - | 3 - 4 | 699 | 60 | |
| AB169508.1 | - | 3 - 4 | 699 | 60 | ||
More Information
Set Membership: |
|

PPP2CA
Genomic Map