Assay Details
| Gene Symbol: B3GALNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005546261.2 | XP_005546318.1 | 5 | 489 | 59 | |
| XM_005546262.2 | XP_005546319.1 | 4 | 720 | 59 | ||
| XM_005546263.2 | XP_005546320.1 | 5 | 460 | 59 | ||
| XM_005546264.2 | XP_005546321.1 | 5 | 629 | 59 | ||
| XM_005546265.2 | XP_005546322.1 | 5 | 563 | 59 | ||
| XM_005546267.2 | XP_005546324.1 | 4 | 394 | 59 | ||
| XM_005546268.2 | XP_005546325.1 | 3 | 294 | 59 | ||
| XM_015445270.1 | XP_015300756.1 | 3 | 634 | 59 | ||
| XM_015445271.1 | XP_015300757.1 | 3 | 448 | 59 | ||
More Information
Set Membership: |
|

B3GALNT1
Genomic Map