Assay Details
| Gene Symbol: TENT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544301.2 | XP_005544358.1 | 1 - 2 | 571 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TENT5B| Gene Symbol: TENT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544301.2 | XP_005544358.1 | 1 - 2 | 571 | 78 | |
Set Membership: |
|