Assay Details
| Gene Symbol: OMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283391.1 | NP_001270320.1 | 2 - 3 | 549 | 89 | |
| XM_005543236.2 | XP_005543293.1 | 2 - 3 | 626 | 89 | ||
| XM_005543237.2 | XP_005543294.1 | 2 - 3 | 608 | 89 | ||
| XM_005543239.2 | XP_005543296.1 | 2 - 3 | 603 | 89 | ||
| XM_015451697.1 | XP_015307183.1 | 2 - 3 | 631 | 89 | ||
| XM_015451702.1 | XP_015307188.1 | 2 - 3 | 629 | 89 | ||
| GenBank mRNA | AB171820.1 | - | 2 - 3 | 549 | 89 | |
More Information
Set Membership: |
|

OMA1
Genomic Map