Assay Details
| Gene Symbol: NACAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549652.2 | XP_005549709.1 | 4 - 5 | 4046 | 76 | |
| XM_015447454.1 | XP_015302940.1 | 5 - 6 | 3929 | 76 | ||
More Information
Set Membership: |
|

NACAD
Genomic Map