Assay Details
| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540579.2 | XP_005540636.1 | 4 - 5 | 2324 | 86 | |
| XM_005540580.2 | XP_005540637.1 | 4 - 5 | 2232 | 86 | ||
| XM_015448908.1 | XP_015304394.1 | 3 - 4 | 1921 | 86 | ||
| XM_015448910.1 | XP_015304396.1 | 3 - 4 | 2021 | 86 | ||
| XM_015448912.1 | XP_015304398.1 | 3 - 4 | 1742 | 86 | ||
More Information
Set Membership: |
|

TMCC2
Genomic Map