Assay Details
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005575603.2 | XP_005575660.1 | 1 - 2 | 590 | 79 | |
XM_015433543.1 | XP_015289029.1 | 1 - 2 | 590 | 79 | ||
XM_015433544.1 | XP_015289030.1 | 1 - 2 | 590 | 79 | ||
XM_015433545.1 | XP_015289031.1 | 1 - 2 | 590 | 79 | ||
XM_015433547.1 | XP_015289033.1 | 1 - 2 | 590 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
