Assay Details
| Gene Symbol: LOC102116121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591047.2 | XP_005591104.1 | 6 - 7 | 832 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102116121| Gene Symbol: LOC102116121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591047.2 | XP_005591104.1 | 6 - 7 | 832 | 87 | |
Set Membership: |
|