Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573610.2 | XP_005573667.1 | 8 - 9 | 1679 | 117 | |
| XM_005573613.2 | XP_005573670.1 | 6 - 7 | 1169 | 117 | ||
| XM_005573615.2 | XP_005573672.1 | 6 - 7 | 3577 | 117 | ||
| XM_005573616.2 | XP_005573673.1 | 6 - 7 | 2629 | 117 | ||
| XM_005573617.2 | XP_005573674.1 | 6 - 7 | 4166 | 117 | ||
| XM_015432442.1 | XP_015287928.1 | 9 - 10 | 1559 | 117 | ||
| XM_015432443.1 | XP_015287929.1 | 7 - 8 | 5316 | 117 | ||
| XM_015432444.1 | XP_015287930.1 | 9 - 10 | 1559 | 117 | ||
| XM_015432445.1 | XP_015287931.1 | 7 - 8 | 9059 | 117 | ||
| XM_015432446.1 | XP_015287932.1 | 7 - 8 | 3616 | 117 | ||
| XM_015432447.1 | XP_015287933.1 | 7 - 8 | 9306 | 117 | ||
| XM_015432448.1 | XP_015287934.1 | 7 - 8 | 4205 | 117 | ||
| XM_015432449.1 | XP_015287935.1 | 7 - 8 | 2672 | 117 | ||
More Information
Set Membership: |
|

ZNF385B
Genomic Map