Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285124.1 | NP_001272053.1 | 8 - 9 | 1032 | 67 | |
| XM_005571217.2 | XP_005571274.1 | 9 - 10 | 1129 | 67 | ||
| GenBank mRNA | AB174355.1 | - | 8 - 9 | 1034 | 67 | |
More Information
Set Membership: |
|

CS
Genomic Map