Assay Details
| Gene Symbol: ETV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588874.2 | XP_005588931.1 | 2 - 3 | 1274 | 99 | |
| XM_005588875.2 | XP_005588932.1 | 3 - 4 | 1169 | 99 | ||
More Information
Set Membership: |
|

ETV2
Genomic Map