Assay Details
Gene Symbol: SMIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005595972.2 | XP_005596029.2 | 2 - 3 | 237 | 77 | |
XM_015449116.1 | XP_015304602.1 | 2 - 3 | 1168 | 77 | ||
XM_015449117.1 | XP_015304603.1 | 2 - 3 | 178 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
