Assay Details
| Gene Symbol: C7H15orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559887.2 | XP_005559944.1 | 1 - 2 | 512 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7H15orf61| Gene Symbol: C7H15orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559887.2 | XP_005559944.1 | 1 - 2 | 512 | 74 | |
Set Membership: |
|