Assay Details
| Gene Symbol: LOC102141746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542535.2 | XP_005542592.1 | 7 - 8 | 1030 | 73 | |
| XM_005542536.2 | XP_005542593.1 | 6 - 7 | 871 | 73 | ||
| XM_015431920.1 | XP_015287406.1 | 7 - 8 | 1742 | 73 | ||
More Information
Set Membership: |
|

LOC102141746
Genomic Map